2,2',5,6'-Tetrachlorbiphenyl (PCB Nr. 53)

Lieferant: EHRENSTORFER

C20005300
EHERC20005300EA 64 EUR
EHERC20005300
2,2',5,6'-Tetrachlorbiphenyl (PCB Nr. 53)
2,2',5,6'-Tetrachlorbiphenyl (PCB Nr. 53)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 41464-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR