2,2',6,6'-Tetrachlorbiphenyl (PCB Nr. 54)

Lieferant: EHRENSTORFER

C20005400
EHERC20005400EA 52 EUR
EHERC20005400
2,2',6,6'-Tetrachlorbiphenyl (PCB Nr. 54)
2,2',6,6'-Tetrachlorbiphenyl (PCB Nr. 54)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 15968-05-5
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR