2,3,3',4'-Tetrachlorbiphenyl (PCB Nr. 56)
Lieferant: EHRENSTORFER
C20005600
EHERC20005600EA
85
EUR
EHERC20005600
2,3,3',4'-Tetrachlorbiphenyl (PCB Nr. 56)
2,3,3',4'-Tetrachlorbiphenyl (PCB Nr. 56)
Formel:
C₁₂H₆Cl₄ Molecular Weight: 291.99 g/mol |
CAS-Nummer:
41464-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...