2,3',4,4'-Tetrachlorbiphenyl (PCB Nr. 66)

Lieferant: EHRENSTORFER

C20006600
EHERC20006600EA 93 EUR
EHERC20006600
2,3',4,4'-Tetrachlorbiphenyl (PCB Nr. 66)
2,3',4,4'-Tetrachlorbiphenyl (PCB Nr. 66)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 32598-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR