2,3',4',5-Tetrachlorbiphenyl (PCB Nr. 70)

Lieferant: EHRENSTORFER

C20007000
EHERC20007000EA 49 EUR
EHERC20007000
2,3',4',5-Tetrachlorbiphenyl (PCB Nr. 70)
2,3',4',5-Tetrachlorbiphenyl (PCB Nr. 70)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 32598-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR