2,3,3',4,5'-Pentachlorbiphenyl (PCB Nr. 108)

Lieferant: EHRENSTORFER

C20010800
EHERC20010800EA 95 EUR
EHERC20010800
2,3,3',4,5'-Pentachlorbiphenyl (PCB Nr. 108)
2,3,3',4,5'-Pentachlorbiphenyl (PCB Nr. 108)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 70362-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR