2,3,3',4',6-Pentachlorbiphenyl (PCB Nr. 110)
Lieferant: EHRENSTORFER
C20011000
EHERC20011000EA
106
EUR
EHERC20011000
2,3,3',4',6-Pentachlorbiphenyl (PCB Nr. 110)
2,3,3',4',6-Pentachlorbiphenyl (PCB Nr. 110)
Formel:
C₁₂H₅Cl₅ Molecular Weight: 326.44 g/mol |
CAS-Nummer:
38380-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...