2,3,3',5,6-Pentachlorbiphenyl (PCB Nr. 112)

Lieferant: EHRENSTORFER

C20011200
EHERC20011200EA 100 EUR
EHERC20011200
2,3,3',5,6-Pentachlorbiphenyl (PCB Nr. 112)
2,3,3',5,6-Pentachlorbiphenyl (PCB Nr. 112)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 74472-36-9
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR