2,2',3,3',4,4'-Hexachlorbiphenyl (PCB Nr. 128)

Lieferant: EHRENSTORFER

C20012800
EHERC20012800EA 87 EUR
EHERC20012800
2,2',3,3',4,4'-Hexachlorbiphenyl (PCB Nr. 128)
2,2',3,3',4,4'-Hexachlorbiphenyl (PCB Nr. 128)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 38380-07-3
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR