2,2',3,3',6,6'-Hexachlorbiphenyl (PCB Nr. 136)

Lieferant: EHRENSTORFER

C20013600
EHERC20013600EA 107 EUR
EHERC20013600
2,2',3,3',6,6'-Hexachlorbiphenyl (PCB Nr. 136)
2,2',3,3',6,6'-Hexachlorbiphenyl (PCB Nr. 136)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 38411-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR