2,2',3,4',5,5'-Hexachlorbiphenyl (PCB Nr. 146)
Lieferant: EHRENSTORFER
C20014600
EHERC20014600EA
228
EUR
EHERC20014600
2,2',3,4',5,5'-Hexachlorbiphenyl (PCB Nr. 146)
2,2',3,4',5,5'-Hexachlorbiphenyl (PCB Nr. 146)
Formel:
C₁₂H₄Cl₆ Molecular Weight: 360.88 g/mol |
CAS-Nummer:
51908-16-8 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...