2,2',4,4',6,6'-Hexachlorbiphenyl (PCB Nr. 155)

Lieferant: EHRENSTORFER

C20015500
EHERC20015500EA 57 EUR
EHERC20015500
2,2',4,4',6,6'-Hexachlorbiphenyl (PCB Nr. 155)
2,2',4,4',6,6'-Hexachlorbiphenyl (PCB Nr. 155)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360.88 g/mol
CAS-Nummer: 33979-03-2
EINECS: 251-773-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR