2,3,3',4,4',5'-Hexachlorbiphenyl (PCB Nr. 157)
Lieferant: EHRENSTORFER
C20015700
EHERC20015700EA
89
EUR
EHERC20015700
2,3,3',4,4',5'-Hexachlorbiphenyl (PCB Nr. 157)
2,3,3',4,4',5'-Hexachlorbiphenyl (PCB Nr. 157)
Formel:
C₁₂H₄Cl₆ Molecular Weight: 360.88 g/mol |
CAS-Nummer:
69782-90-7 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...