2,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 167)
Lieferant: EHRENSTORFER
C20016700
EHERC20016700EA
98
EUR
EHERC20016700
2,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 167)
2,3',4,4',5,5'-Hexachlorbiphenyl (PCB Nr. 167)
Formel:
C₁₂H₄Cl₆ Molecular Weight: 360.88 g/mol Siedepunkt: 383 Schmelzpunkt: 125…127 °C |
CAS-Nummer:
52663-72-6 UN: 3432 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...