2,2',3,3',4,4',5-Heptachlorbiphenyl (PCB Nr. 170)

Lieferant: EHRENSTORFER

C20017000
EHERC20017000EA 89 EUR
EHERC20017000
2,2',3,3',4,4',5-Heptachlorbiphenyl (PCB Nr. 170)
2,2',3,3',4,4',5-Heptachlorbiphenyl (PCB Nr. 170)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 35065-30-6
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR