Decachlorbiphenyl (PCB Nr. 209)

Lieferant: EHRENSTORFER

Synonyme: 2,2',3,3',4,4',5,5',6,6'-Decachlorbiphenyl, PCB Nr. 209, Pentachlor(pentachlorphenyl)benzol, 2,2',3,3',4,4',5,5',6,6'-PCB

C20020900
EHERC20020900EA 39 EUR
EHERC20020900
Decachlorbiphenyl (PCB Nr. 209)
Decachlorbiphenyl (PCB Nr. 209)
Formel: C₁₂Cl₁₀
Molecular Weight: 498.66 g/mol
CAS-Nummer: 2051-24-3
EINECS: 218-115-1
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR