Octachlornaphthalin
Lieferant: EHRENSTORFER
|
Achtung
|
C20425800
EHERC20425800EA
52
EUR
EHERC20425800
Octachlornaphthalin
Octachlornaphthalin
Formel:
C₁₀Cl₈ Molecular Weight: 403.73 g/mol |
CAS-Nummer:
2234-13-1 EINECS: 218-778-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...