Acenaphthen-[D10]
Lieferant: EHRENSTORFER
|
|
Synonyme:
Acenaphthene-[²H₁₀]
C20505100
EHERC20505100EA
43
EUR
EHERC20505100
Acenaphthen-[D10]
Acenaphthen-[D10]
Formel:
C₁₂D₁₀ Molecular Weight: 164,13 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
15067-26-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...