Benzo[j]fluoranthen
Lieferant: EHRENSTORFER
C20575000
EHERC20575000EA
101
EUR
EHERC20575000
Benzo[j]fluoranthen
Benzo[j]fluoranthen
Formel:
C₂₀H₁₂ |
CAS-Nummer:
205-82-3 EINECS: 205-910-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...