Benzo[k]fluoranthen

Lieferant: EHRENSTORFER

C20580000
EHERC20580000EA 70 EUR
EHERC20580000
Benzo[k]fluoranthen
Benzo[k]fluoranthen
Formel: C₂₀H₁₂
MDL: MFCD00046287
CAS-Nummer: 207-08-9
EINECS: 205-916-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR