Benzo[a]pyren-[D12]

Lieferant: EHRENSTORFER
Gefahr

C20635100
EHERC20635100EA 403 EUR
EHERC20635100
Benzo[a]pyren-[D12]
Benzo[a]pyren-[D12]
Formel: C₂₀D₁₂
CAS-Nummer: 63466-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR