Chrysen-[D12]
Lieferant: EHRENSTORFER
C20670100
EHERC20670100EA
211
EUR
EHERC20670100
Chrysen-[D12]
Chrysen-[D12]
Formel:
C₁₈D₁₂ |
CAS-Nummer:
1719-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...