Fluoranthen

Lieferant: EHRENSTORFER
Achtung

C20795000
EHERC20795000EA 42 EUR
EHERC20795000
Fluoranthen
Fluoranthene
Formel: C₁₆H₁₀
Molecular Weight: 202,26 g/mol
MDL: MFCD00001184
CAS-Nummer: 206-44-0
EINECS: 205-912-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR