Fluoranthen-[D10]

Lieferant: EHRENSTORFER
Achtung

Synonyme: Fluoranthene-[²H₁₀]

C20795100
EHERC20795100EA 310 EUR
EHERC20795100
Fluoranthen-[D10]
Fluoranthen-[D10]
Formel: C₁₆D₁₀
CAS-Nummer: 93951-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR