5-Methylbenzo[b]thiophen
Lieferant: EHRENSTORFER
C20846000
EHERC20846000EA
64
EUR
EHERC20846000
5-Methylbenzo[b]thiophen
5-Methylbenzo[b]thiophen
Formel:
C₉H₈S Molecular Weight: 148.23 g/mol |
MDL:
MFCD00052509 CAS-Nummer: 14315-14-1 EINECS: 238-256-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...