2-Brombiphenyl (PBB Nr. 1)

Lieferant: EHRENSTORFER

Synonyme: 1-Brom-2-phenylbenzol, PBB Nr. 1

C21000100
EHERC21000100EA 38 EUR
EHERC21000100
2-Brombiphenyl (PBB Nr. 1)
2-Brombiphenyl (PBB Nr. 1)
Formel: C₁₂H₉Br
Molecular Weight: 233,11 g/mol
Siedepunkt: 297 °C (1013 hPa)
Schmelzpunkt: 2 °C
Dichte: 1,2766 g/cm³ (20 °C)
MDL: MFCD00000065
CAS-Nummer: 2052-07-5
EINECS: 218-141-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR