2,4,5-Tribrombiphenyl (PBB Nr. 29)

Lieferant: EHRENSTORFER

C21002900
EHERC21002900EA 62 EUR
EHERC21002900
2,4,5-Tribrombiphenyl (PBB Nr. 29)
2,4,5-Tribrombiphenyl (PBB Nr. 29)
Formel: C₁₂H₇Br₃
CAS-Nummer: 115245-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR