2-Ethylhexylacrylat

Lieferant: EHRENSTORFER

Synonyme: Propensäure(2-ethylhexyl)ester, Acrylsäure(2-ethylhexyl)ester

CA10045390
EHERCA10045390EA 34 EUR
EHERCA10045390
2-Ethylhexylacrylat
2-Ethylhexylacrylat
Formel: C₁₁H₂₀O₂
Molecular Weight: 184.28 g/mol
MDL: MFCD00009495
CAS-Nummer: 103-11-7
EINECS: 203-080-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR