1-Butyl-2-pyrrolidone
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
N-Butyl-2-azacyclopentanone, 1-Butylpyrrolidin-2-one
CA10931640
EHERCA10931640EA
85.3
EUR
EHERCA10931640
1-Butyl-2-pyrrolidone
1-Butyl-2-pyrrolidone
Formel:
C₈H₁₅NO Molecular Weight: 141,21 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00020876 CAS-Nummer: 3470-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...