Disulfoton
Lieferant: EHRENSTORFER
CA12980000
EHERCA12980000EA
80
EUR
EHERCA12980000
Disulfoton
Disulfoton
Formel:
C₈H₁₉O₂PS₃ |
CAS-Nummer:
298-04-4 EINECS: 206-054-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...