Ethephon hydroxy
Lieferant: EHRENSTORFER
|
Gefahr
|
Synonyme:
(2-Hydroxyethyl)phosphonic acid
CA13230200
EHERCA13230200EA
465
EUR
EHERCA13230200
Ethephon hydroxy
Ethephon hydroxy
Formel:
C₂H₇O₄P Molecular Weight: 126,0483 g/mol |
MDL:
MFCD00043865 CAS-Nummer: 22987-21-9 UN: 3265 ADR: , |
Spezifikation Testergebnisse
Form | Viscous |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...