Ethoxyquin

Lieferant: EHRENSTORFER

Synonyme: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

CA13310000
EHERCA13310000EA 57 EUR
EHERCA13310000
Ethoxyquin
Ethoxyquin
Formel: C₁₄H₁₉NO
MDL: MFCD00023883
CAS-Nummer: 91-53-2
EINECS: 202-075-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR