(±)-Methyl-2-methylbutyrat

Lieferant: EHRENSTORFER

Synonyme: Methyl2-methylbutyrate

CA15084850
EHERCA15084850EA 43 EUR
EHERCA15084850
(±)-Methyl-2-methylbutyrat
(±)-Methyl-2-methylbutyrat
Formel: C₆H₁₂O₂
Molecular Weight: 116,16 g/mol
MDL: MFCD00009335
CAS-Nummer: 868-57-5
EINECS: 212-778-0
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR