2,6-Toluoldiisocyanat
Lieferant: EHRENSTORFER
Synonyme:
2,6-TDI, 2-Methyl-m-phenylendiisocyanat, Tolylen-2,6-diisocyanat
CA17594600
EHERCA17594600EA
74
EUR
EHERCA17594600
2,6-Toluoldiisocyanat
2,6-Toluoldiisocyanat
Formel:
C₉H₆N₂O₂ Molecular Weight: 174.16 |
MDL:
MFCD00002010 CAS-Nummer: 91-08-7 EINECS: 202-039-0 UN: 2078 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...