Vitamin A acetat
Lieferant: EHRENSTORFER
Synonyme:
alle-trans-Retinylacetat
CA17923820
EHERCA17923820EA
102
EUR
EHERCA17923820
Vitamin A acetat
alle-trans-Retinylacetat
Formel:
C₂₂H₃₂O₂ Molecular Weight: 328,49 g/mol |
MDL:
MFCD00019413 CAS-Nummer: 127-47-9 EINECS: 204-844-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...