O15-Palmitoylretinol

Lieferant: EHRENSTORFER
Achtung

Synonyme: Retinyl palmitate, Retinol palmitate, alle-trans-Retinylpalmitat

CA17923840
EHERCA17923840EA 63 EUR
EHERCA17923840
O15-Palmitoylretinol
Vitamin A palmitate
Formel: C₃₆H₆₀O₂
Molecular Weight: 524,87 g/mol
MDL: MFCD00019414
CAS-Nummer: 79-81-2
EINECS: 201-228-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR