DL-α-Tocopherol

Lieferant: EHRENSTORFER

CA17924300
EHERCA17924300EA 51 EUR
EHERCA17924300
DL-α-Tocopherol
DL-α-Tocopherol
Formel: C₂₉H₅₀O₂
MDL: MFCD00072051
CAS-Nummer: 10191-41-0
EINECS: 233-466-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR