Organic reference standard, Acetamiprid in Ethylacetate, Dr. Ehrenstorfer
Lieferant: EHRENSTORFER
L10013000EA
EHERL10013000EAEA
64
EUR
EHERL10013000EA
Organic reference standard, Acetamiprid in Ethylacetate, Dr. Ehrenstorfer
Standards
Organische Standards
- Solubility in water at 25 °C: 100 g/l
- 10 µg/ml in ethyl acetate