Pestizid- Referenzstandard, Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat) 10 µg/ml in Acetonitril
Lieferant: EHRENSTORFER
L17270000AL
EHERL17270000ALEA 49 EUR
EHERL17270000AL
Pestizid- Referenzstandard, Terbufos (S-tert-Butylthiomethyl-O,O-diethyldithiophosphat) 10 µg/ml in Acetonitril