Organic reference standard, Geosmin, Dr. Ehrenstorfer
Lieferant: EHRENSTORFER
LA14005000ME
EHERLA14005000MEEA
84
EUR
EHERLA14005000ME
Organic reference standard, Geosmin, Dr. Ehrenstorfer
Standards
Organische Standards
This standard solution is designed for calibration or recalibration of chromatographic systems for the determination of the specified chemical compounds concerning identity and qualification. It can be also used as reference material for interlaboratory studies to validate analytical procedures.
A serial numbered certificate is provided from an ISO 17025 calibration to indicate the instrument standard.
Standards: Accredited to ISO/IEC 17025.