Universal vial caps
Lieferant: EKSIGENT
A62250
EKSIA62250EA 148 EUR
EKSIA62250
Universal vial caps
Verschlüsse Chromatographieampullen-Verschlüsse
Universal caps are used in combination with universal vials.
Universal vial caps are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.