Low pH SDS Sample Buffer
Lieferant: EKSIGENT
C44807
EKSIC44807EA 197 EUR
EKSIC44807
Low pH SDS Sample Buffer
Puffer Elektrophorese-Puffer Beladungspuffer
SCIEX Low pH SDS Sample Buffer, 140 mL (100 mM Tris-HCl, pH 6.8, 1% SDS) allows you to use SCIEX’s Gold Standard CE-SDS with a low pH SDS sample buffer. Use of this buffer can increase non-reduced purity by decreasing method induced fragmentation. Less variability and lower RSDs means less troubleshooting and faster method development.