Cole-Parmer® HM-200 Heating Mantles
Lieferant: Cole-Parmer
36002-10 36002-14 36002-18 36002-02 36002-06 36002-30 36002-22 36002-26
ELET36002-10EA 837 EUR
ELET36002-10 ELET36002-14 ELET36002-18 ELET36002-02 ELET36002-06 ELET36002-30 ELET36002-22 ELET36002-26
Cole-Parmer® HM-200 Heating Mantles
Heizgeräte Heizhauben
Thermal insulating cartridge provides maximum heat transfer.
  • Chemical resistant polypropylene housing is 'safe to the touch'
Quickly and safely heat round-bottom flasks to 450 °C (842 °F). These simple-to-use heating mantles feature a built-in power controller with dial markings from 1 to 10 for repeatability. An amber coloured 'heater on' light and a clear-coloured 'power on' light indicate when the heating mantle is in use. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer andMore Product Information