Water purification systems, MEDICA® 7/15
Lieferant: ELGA LabWater
MR007BPM1 MR007XXM1 MR015BPM1 MR015XXM1 ME007BPM1 ME007XXM1 ME015BPM1 ME015XXM1
ELGAMR007BPM1EA 9459 EUR
ELGAMR007BPM1 ELGAMR007XXM1 ELGAMR015BPM1 ELGAMR015XXM1 ELGAME007BPM1 ELGAME007XXM1 ELGAME015BPM1 ELGAME015XXM1
Water purification systems, MEDICA® 7/15
Wasseraufbereitungssysteme Reinstwassersysteme
Medica® 7/15 water systems are high performance water purification systems that directly feed clinical diagnostic analysers.
  • Available in recirculating models (Medica® R) or standard systems (Medica® S)
  • Flow rates from 7 to 15 L/h
  • Models with booster pump are also available
Medica® R 7/15 are compact, high performance water purification systems that directly feed Clinical diagnostic analysers with up to 1,8 L/min of CLSI type 1 water with productivity of 7 or 15 L/h. Innovative technology, combined with exceptional build quality, makes the MEDICA® R 7/More Product Information