Magnesiumperchlorat
Lieferant: ELTRA
|
Achtung
|
Synonyme:
Anhydron
90200
ELTA90200EA
116
EUR
ELTA90200
Magnesiumperchlorat
Magnesiumperchlorat
Formel:
Cl₂MgO₈ Molecular Weight: 223,21 g/mol Schmelzpunkt: 251 °C Dichte: 2,21 g/cm³ (20 °C) |
MDL:
MFCD00011102 CAS-Nummer: 10034-81-8 EINECS: 233-108-3 UN: 1475 ADR: 5.1,II Merck Index: 13,05702 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...