Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
EM0200-1KT
EM0200-1KTEA 396 EUR
EM0200-1KT
Osteo-Bed Bone embedding kit, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Suitable for use with large and small mineralized (undecalcified) bone sections. Yields clear, hard blocks for cutting sections. Not water-soluble.
  • Quality Level 200
Kit contains:
Osteo-Bed Bone Embedding Resin A (O8514) 900 ml
Osteo-Bed Bone Embedding Catalyst (O8764) 2 x 12 g