Insulinspritzen, BD Micro-Fine™
Lieferant: EMBECTA
324892 324891 324826 324825 324827 324824
EMBE324892EA 133 EUR
EMBE324892 EMBE324891 EMBE324826 EMBE324825 EMBE324827 EMBE324824
Insulinspritzen, BD Micro-Fine™
Spritzen Spritzen, medizinisch
Micro-Fine™ is a single-use syringe for diabetes. It offers a more comfortable and convenient way to inject insulin with three different volumes.
  • Made from surgical stainless steel
  • Precision and comfort
  • Optimum shape and sharpness of the needle tip and bevel
  • Electro polished needle shaft
The white and orange protective cap conforms with sterility and a minimal dead space reduces the risk of air bubbles and prevents waste of insulin.