3-Cyan-2-methylanilin

Lieferant: SIA ENAMINE

EN300-112929
ENAMEN300-112929EA 0 EUR
ENAMEN300-112929
3-Cyan-2-methylanilin
3-Cyan-2-methylanilin
Formel: C₈H₈N₂
CAS-Nummer: 69022-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR