5-Chlor-2-methyl-7H-isoxazolo[2,3-a]pyrimidin-7-on

Lieferant: SIA ENAMINE

EN300-14372.1G
ENAMEN300-14372.1GEA 0 EUR
ENAMEN300-14372.1G
5-Chlor-2-methyl-7H-isoxazolo[2,3-a]pyrimidin-7-on
5-Chlor-2-methyl-7H-isoxazolo[2,3-a]pyrimidin-7-on
Formel: C₇H₅ClN₂O₂
MDL: MFCD07366359
CAS-Nummer: 278614-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR