5'-Chlor-2'-methoxyacetophenon

Lieferant: SIA ENAMINE

Synonyme: 1-(5-Chloro-2-methoxyphenyl)ethanone

EN300-14420.5G
ENAMEN300-14420.5GEA 0 EUR
ENAMEN300-14420.5G
5'-Chlor-2'-methoxyacetophenon
5'-Chlor-2'-methoxyacetophenon
Formel: C₉H₉ClO₂
Molecular Weight: 184,6196 g/mol
MDL: MFCD00174339
CAS-Nummer: 6342-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR